4-Chloro-1-methylpiperidine hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C1085-25G C1085-100G
TCIAC1085-25GEA 337 DKK
TCIAC1085-25G TCIAC1085-100G
4-Chloro-1-methylpiperidine hydrochloride ≥98.0% (by titrimetric analysis)
4-Chloro-1-methylpiperidine hydrochloride
Formel: C₆H₁₃Cl₂N
Molekylvægt: 170.08 g/mol
MDL Number: MFCD00012834
CAS-nummer: 5382-23-0
EINECS: 226-375-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eee9d54e1d010e43aaa64f614ebc65f5d89cd07f&vid=eee9d54e1d010e43aaa64f614ebc65f5d89cd07f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 161.0 to 166.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR