2-(4-Methylphenyl)ethanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M2298-25G M2298-5G
TCIAM2298-25GEA 1870 DKK
TCIAM2298-25G TCIAM2298-5G
2-(4-Methylphenyl)ethanol ≥98.0% (by GC)
2-(4-Methylphenyl)ethanol
Formel: C₉H₁₂O
Kogepunkt: 115°C
Flammepunkt: 107°C
MDL Number: MFCD00002904
CAS-nummer: 699-02-5
EINECS: 211-824-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a8ef9394e7a624b98daadc1fd128bb00416c935&vid=5a8ef9394e7a624b98daadc1fd128bb00416c935&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR