2-(3-Methylphenyl)ethanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M1376-25G M1376-5G
TCIAM1376-25GEA 2750 DKK
TCIAM1376-25G TCIAM1376-5G
2-(3-Methylphenyl)ethanol ≥98.0% (by GC)
2-(3-Methylphenyl)ethanol
Formel: C₉H₁₂O
Kogepunkt: 243°C
Flammepunkt: 109°C
MDL Number: MFCD00002896
CAS-nummer: 1875-89-4
EINECS: 217-508-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51401295dcda51fe57f48c55b48241e9f3bd59ea&vid=51401295dcda51fe57f48c55b48241e9f3bd59ea&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR