Dioctadecyl ether ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
O0016-25G
O0016-250G
TCIAO0016-25GEA
846
DKK
TCIAO0016-25G
TCIAO0016-250G
Dioctadecyl ether ≥95.0% (by GC)
Dioctadecyl ether
Formel:
C₃₆H₇₄O Kogepunkt: 131°C |
MDL Number:
MFCD00048498 CAS-nummer: 6297-03-6 EINECS: 228-567-1 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 61.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...