Gold Palladium (60:40) (w/w) ≥99.9% (metals basis), wire, annealed, Ø 0.2 mm (0.008 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41206.BU
41206.BW
41206.G1
41206.BUEA
1350
DKK
41206.BU
41206.BW
41206.G1
Gold Palladium (60:40) (w/w) ≥99.9% (metals basis), wire, annealed, Ø 0.2 mm (0.008 in)
Gold Palladium
Purity: 99.9%
Molecular Formula: Au:Pd; 60:40 wt%
Physical Form: Temper: soft est .154g/ft
MDL No.: MFCD00003436