4-Bromoisatin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B2423-25G B2423-5G
TCIAB2423-25GEA 731 DKK
TCIAB2423-25G TCIAB2423-5G
4-Bromoisatin ≥98.0% (by GC, titration analysis)
4-Bromoisatin
Formel: C₈H₄BrNO₂
Smeltepunkt: 271°C
MDL Number: MFCD03618555
CAS-nummer: 20780-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73021bac73fffd6b4de887d9383bfb04a8f8e7f1&vid=73021bac73fffd6b4de887d9383bfb04a8f8e7f1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR