Benzyl phenyl ether ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B0433-25G B0433-500G
TCIAB0433-25GEA 403 DKK
TCIAB0433-25G TCIAB0433-500G
Benzyl phenyl ether ≥98.0% (by GC)
Benzyl phenyl ether
Formel: C₁₃H₁₂O
Kogepunkt: 288°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00020660
CAS-nummer: 946-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a445565700b8ef372ead20aea9ae30f09e089dac&vid=a445565700b8ef372ead20aea9ae30f09e089dac&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR