Potassium (4-pyridyl)trifluoroborate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Potassium 4-Pyridyltrifluoroborate

P1684-1G P1684-5G
TCIAP1684-1GEA 509 DKK
TCIAP1684-1G TCIAP1684-5G
Potassium (4-pyridyl)trifluoroborate ≥98.0% (by titrimetric analysis)
Potassium (4-pyridyl)trifluoroborate
Formel: C₅H₄BF₃KN
MDL Number: MFCD09038543
CAS-nummer: 1111732-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bffea16bbdd07358f693ad86404140e2e876561a&vid=bffea16bbdd07358f693ad86404140e2e876561a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR