Biphenyl-4-sulphonyl chloride ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Biphenyl-4-sulphonyl Chloride
B2199-25G
B2199-5G
TCIAB2199-25GEA
1240
DKK
TCIAB2199-25G
TCIAB2199-5G
Biphenyl-4-sulphonyl chloride ≥98.0% (by GC)
Biphenyl-4-sulphonyl chloride
Formel:
C₁₂H₉ClO₂S |
MDL Number:
MFCD00078658 CAS-nummer: 1623-93-4 EINECS: 216-609-1 UN nummer: 3261 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...