Methyl-5-methylsalicylate ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 2-hydroxy-5-methylbenzoate

M1353-25G M1353-5G
TCIAM1353-25GEA 1700 DKK
TCIAM1353-25G TCIAM1353-5G
Methyl-5-methylsalicylate ≥96.0% (by GC)
Methyl-5-methylsalicylate
Formel: C₉H₁₀O₃
Molekylvægt: 166.1700 g/mol
Kogepunkt: 242°C
Flammepunkt: 108°C
MDL Number: MFCD00016485
CAS-nummer: 22717-57-3
EINECS: 245-173-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eebaad9c91591d2703e4e07b4417a1f42a899f0&vid=8eebaad9c91591d2703e4e07b4417a1f42a899f0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR