Thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T2193-25G T2193-5G
TCIAT2193-25GEA 2040 DKK
TCIAT2193-25G TCIAT2193-5G
Thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
Thiomorpholine-1,1-dioxide
Formel: C₄H₉NO₂S
Kogepunkt: 155°C
MDL Number: MFCD05861623
CAS-nummer: 39093-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed643803038fb4c951da2e601b0e8ccbe31f9f4e&vid=ed643803038fb4c951da2e601b0e8ccbe31f9f4e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 68.0 to 71.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR