4-(Dimethylamino)-2-methoxybenzaldehyde 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B23598.03
B23598.06
B23598.03EA
535
DKK
B23598.03
B23598.06
4-(Dimethylamino)-2-methoxybenzaldehyde 98%
4-(Dimethylamino)-2-methoxybenzaldehyde
Purity: 98%
CAS No.: 84562-48-1
Molecular Formula: C10H13NO2
Formula Weight: 179.22
Melting Point: 58-60°
Beilstein Reference: 2719181
Formel:
C₁₀H₁₃NO₂ Molekylvægt: 179.22 g/mol Smeltepunkt: 58…60 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00151814 CAS-nummer: 84562-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...