N-Boc-D-valine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Boc-D-Val-OH
B2991-25G
B2991-5G
TCIAB2991-25GEA
813
DKK
TCIAB2991-25G
TCIAB2991-5G
N-Boc-D-valine ≥98.0% (by HPLC, titration analysis)
N-Boc-D-valine
Formel:
C₁₀H₁₉NO₄ Molekylvægt: 217.3 g/mol |
MDL Number:
MFCD00038282 CAS-nummer: 22838-58-0 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Specific rotation [a]20/D | +5.0 to +8.0 deg (C=1, AcOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...