1,3-Propanedisulphonic acid disodium salt 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Sodium propane-1,3-disulphonate
B20092.06
B20092.14
B20092.06EA
477
DKK
B20092.06
B20092.14
1,3-Propanedisulphonic acid disodium salt 99%
1,3-Propanedisulphonic acid disodium salt
Purity: 99%
CAS No.: 36589-58-9
Molecular Formula: NaO3S(CH2)3SO3Na
Formula Weight: 248.19
Melting Point:
Formel:
C₃H₆Na₂O₆S₂ Molekylvægt: 248.19 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00191483 CAS-nummer: 36589-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...