(Chloromethyl)dimethylphenylsilane ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(Chloromethyl)dimethyl(phenyl)silane
C1419-25G
C1419-5G
TCIAC1419-25GEA
1480
DKK
TCIAC1419-25G
TCIAC1419-5G
(Chloromethyl)dimethylphenylsilane ≥97.0% (by GC)
(Chloromethyl)dimethylphenylsilane
Formel:
C₉H₁₃ClSi Kogepunkt: 225°C Flammepunkt: 91°C |
MDL Number:
MFCD00040858 CAS-nummer: 1833-51-8 EINECS: 217-392-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...