(R)-3-(Aminomethyl)-1-boc-piperidine 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-1-BOC-3-(Aminomethyl)piperidine

H52561.MD H52561.03 H52561.06
H52561.MDEA 614 DKK
H52561.MD H52561.03 H52561.06
(R)-3-(Aminomethyl)-1-boc-piperidine 97%
(R)-3-(Aminomethyl)-1-boc-piperidine

Purity: 97% CAS No.: 140645-23-4 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 MDL No.: MFCD03839876

Formel: C₁₁H₂₂N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03839876
CAS-nummer: 140645-23-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR