3-(Trifluoromethoxy)cinnamic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-(3-(Trifluoromethoxy)phenyl)acrylic acid

T1780-25G T1780-5G
TCIAT1780-25GEA 2780 DKK
TCIAT1780-25G TCIAT1780-5G
3-(Trifluoromethoxy)cinnamic acid ≥98.0% (by titrimetric analysis)
3-(Trifluoromethoxy)cinnamic acid
Formel: C₁₀H₇F₃O₃
Molekylvægt: 232.16 g/mol
MDL Number: MFCD00066337
CAS-nummer: 168833-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed9be395fd3315231f3d640344c651217b452a6a&vid=ed9be395fd3315231f3d640344c651217b452a6a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 94.0 to 97.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR