4-(Dimethylamino)cinnamic acid 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-(Dimethylamino)cinnamic Acid

B21549.03 B21549.06
B21549.03EA 809 DKK
B21549.03 B21549.06
4-(Dimethylamino)cinnamic acid 99%
4-(Dimethylamino)cinnamic acid

Purity: 99% CAS No.: 1552-96-1 Molecular Formula: C11H13NO2 Formula Weight: 191.23 Melting Point: ca 222° dec. Beilstein Reference: 1368533

Formel: C₁₁H₁₃NO₂
Molekylvægt: 191.23 g/mol
Smeltepunkt: ∼ 222 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004397
CAS-nummer: 1552-96-1
EINECS: 216-299-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR