Isonipecotamide ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A13442.09 A13442.18 A13442.30
A13442.09EA 292 DKK
A13442.09 A13442.18 A13442.30
Isonipecotamide ≥98%
Isonipecotamide

Purity: 98% CAS No.: 39546-32-2 Molecular Formula: C6H12N2O Formula Weight: 128.18 Melting Point: 148-149° Beilstein Reference: 112554

Formel: C₆H₁₂N₂O
Smeltepunkt: 148…149 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00038012
CAS-nummer: 39546-32-2
EINECS: 254-501-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR