Nipecotamide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

N0097-25G N0097-5G
TCIAN0097-25GEA 591 DKK
TCIAN0097-25G TCIAN0097-5G
Nipecotamide ≥98.0% (by titrimetric analysis)
Nipecotamide
Formel: C₆H₁₂N₂O
MDL Number: MFCD00005993
CAS-nummer: 4138-26-5
EINECS: 223-962-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a50db96570aceaa80ab7e94fa7d425c4f1711c07&vid=a50db96570aceaa80ab7e94fa7d425c4f1711c07&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 108.0 to 113.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR