Tetrazolium violet ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T0174-1G T0174-10G
TCIAT0174-1GEA 288 DKK
TCIAT0174-1G TCIAT0174-10G
Tetrazolium violet ≥98.0% (by titrimetric analysis)
Tetrazolium violet
Formel: C₂₃H₁₇ClN₄
MDL Number: MFCD00011875
CAS-nummer: 1719-71-7
EINECS: 217-008-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=510e1faf3bd65a1c653ad1243e7354500c93c7a8&vid=510e1faf3bd65a1c653ad1243e7354500c93c7a8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in hot Water faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR