Bathocuproin sulphonate disodium salt hydrate ≥97%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Bathocuproindisulphonic disodium salt hydrate
B22550.03 B22550.06 B22550.MD
B22550.03EA 1130 DKK
B22550.03 B22550.06 B22550.MD
Bathocuproin sulphonate disodium salt hydrate ≥97%
Bathocuproin sulphonate disodium salt hydrate
Purity: 97% CAS No.: 52698-84-7 Molecular Formula: C26H18N2Na2O6S2.xH2O Formula Weight: 564.55(anhy) Melting Point: 300° Beilstein Reference: 9033637
Formel: C₂₆H₁₈N₂Na₂O₆S₂·H₂O
Molekylvægt: 564.55 g/mol (anhy)
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03012947
CAS-nummer: 52698-84-7
EINECS: 258-111-7