4-(Benzyloxy)aniline hydrochloride ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-Benzyloxyanilinium Chloride, 4-Aminophenyl benzyl ether hydrochloride, p-Benzyloxyaniline hydrochloride
A16856.14
A16856.22
A16856.14EA
480
DKK
A16856.14
A16856.22
4-(Benzyloxy)aniline hydrochloride ≥98%
4-(Benzyloxy)aniline hydrochloride
Purity: 98%
CAS No.: 51388-20-6
Molecular Formula: C13H13NO.HCl
Formula Weight: 235.72
Melting Point: ca 225° dec.
Beilstein Reference: 3633307
Formel:
C₁₃H₁₄ClNO Smeltepunkt: ∼ 225 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012995 CAS-nummer: 51388-20-6 EINECS: 257-170-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...