4-(Benzyloxy)aniline hydrochloride 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Benzyloxyanilinium Chloride, 4-Aminophenyl benzyl ether hydrochloride, p-Benzyloxyaniline hydrochloride
152290250
152291000
ACRO152290250EA
299
DKK
ACRO152290250
ACRO152291000
4-(Benzyloxy)aniline hydrochloride 98%
4-(Benzyloxy)aniline hydrochloride
Formel:
C₁₃H₁₄ClNO Smeltepunkt: 228 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012995 CAS-nummer: 51388-20-6 EINECS: 257-170-6 |
Specifikationer Testresultater
Appearance | Brown Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...