Dibutylbis(2,4-pentanedionato-O,O')tin 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Di-n-butyltinbis(acetylacetonate), Di-n-butyltin bis(2,4-pentanedionate), (OC-6-11)-Dibutylbis(2,4-pentanedionato-κO2,κO4)Tin

B23921.0B B23921.18 B23921.30
B23921.0BEA 3560 DKK
B23921.0B B23921.18 B23921.30
Dibutylbis(2,4-pentanedionato-O,O')tin 95%
Dibutylbis(2,4-pentanedionato-O,O')tin

Purity: 95% CAS No.: 22673-19-4 Molecular Formula: C18H32O4Sn Formula Weight: 431.16 Flash Point: 40°(104°F) Refractive Index: 1.5150 Odour: Weak Melting Point: 26° Boiling Point: 150° UN No.: 1992 MDL No.: MFCD00077994

Formel: C₁₈H₃₂O₄Sn
Kogepunkt: 150 °C
Smeltepunkt: 26 °C
Massefylde: 1,22
Flammepunkt: 40 °C (104 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00077994
CAS-nummer: 22673-19-4
EINECS: 245-152-0
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR