3-Fluoro-4-iodoaniline 98

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H61427.03 H61427.06
H61427.03EA 961 DKK
H61427.03 H61427.06
3-Fluoro-4-iodoaniline 98
3-Fluoro-4-iodoaniline
Formel: C₆H₅FIN
Molekylvægt: 237.01 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07774187
CAS-nummer: 656-66-6
UN nummer: 2811
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR