2-Fluoro-m-toluidine ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Fluoro-3-methylaniline, 3-Amino-2-fluorotoluene

H61143.14 H61143.22
H61143.14EA 2050 DKK
H61143.14 H61143.22
2-Fluoro-m-toluidine ≥98%
2-Fluoro-m-toluidine

Purity: 98% CAS No.: 1978-33-2 Molecular Formula: C7H8FN Formula Weight: 125.14 Refractive Index: 1.5360 Boiling Point: 87°/12mm UN No.: 2810 MDL No.: MFCD06410915

Formel: C₇H₈FN
Molekylvægt: 125.14 g/mol
Kogepunkt: 87 °C (12 mmHg)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06410915
CAS-nummer: 1978-33-2
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR