Sterilising station, SteriMax
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KL88.1 LH89.1
ROTHKL88.1EA 5224.88 DKK
ROTHKL88.1 ROTHLH89.1
Sterilising station, SteriMax
Varmeapparater Gasbrændere
Stainless steel sterilising station for sterilising inoculating loops, needles and instruments. Ideal for laboratories and safety workbenches where gas and open fires cannot be used. Simply hold the inoculating loop in the quartz glass sterilising tube to sterilise. The loop is sterile after approx. 5 to 7 s.
  • Sterilising temperature 900 to 950 °C
  • Residual heat display warns against high temperatures
  • Angle can be adjusted up to seven times
  • Integrated splash protection
  • Trays for three instruments