Microplates, 96-well, non binding
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
650901_40 655901_40 655900_40 655906_40 655903_40 655904_40 651901_40
738-0165EA 1900 DKK
738-0165 738-0170 738-0169 738-0173 738-0171 738-0172 738-0166
Microplates, 96-well, non binding
Mikroplader HTS mikroplader
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.