Racks for upright freezers, with sliding shelves and depth 670 mm, Comfort racks
Leverandør: TENAK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TE24252-S TE24420-S TE24286-S TE24310-S
479-0998EA 1530 DKK
479-0998 479-1847 479-0996 479-0997
Racks for upright freezers, with sliding shelves and depth 670 mm, Comfort racks
Stativer Stativer til Kryobokse
Stainless steel.
  • Racks for four different box heights are available
  • Fit all standard cryoboxes
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility.
Note: Measure the inside height and depth of your freezer to ensure the right choice of rack.
Leveringsinformation: Delivered without cryoboxes.