Drum taps, brass
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2798 2799 2800
MSGA2798EA 521 DKK
MSGA2798 MSGA2799 MSGA2800
Drum taps, brass
Vandhaner og håndvaske Vandhaner
Brass taps for drums with screw thread.
  • Self closing
  • Flame guard
  • PTFE seal
Bestillingsoplysninger: MSGA2799 includes a trailer fitting for decanting vessels.