Capillary tubing, PEEK
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37005-50 37007 37007-100 37010-100 37003
555-5140EA 258 DKK
555-5140 555-5139 555-5138 555-5136 555-5202
Capillary tubing, PEEK
Slanger Kromatografirør
PEEK tubing.
  • Broad chemical compatibility
  • Biocompatible
  • Resistant to most organic solvents
  • Available in a range of internal diameters
This tubing is designed to work in most HPLC applications, particularly reversed phase with aqueous mobile phases.