VWR® Cryolab Q Path, Freezing Aerosol
Leverandør: VWR Q-Path Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
00528200.EA 1050 DKK
00528200. 13328202.
VWR® Cryolab Q Path, Freezing Aerosol
Farvestoffer Mikroskopi og farvningsreagenser
Specially used for freezing of surgical parts before the cutting process with cryostat or with a freezing microtome.
  • Storage temperature: Ambient
  • UN 1950