ESD safety shoes, slip-on, HACCP, X-Light, 7131028/7131029
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7131028/35 7131028/36 7131028/37 7131028/38 7131028/39 7131028/40 7131028/41 7131028/42 7131028/43 7131028/44 7131028/45 7131028/46 7131028/47 7131028/48 7131029/35 7131029/36 7131029/37 7131029/38 7131029/39 7131029/40 7131029/41 7131029/42 7131029/43 7131029/44 7131029/45 7131029/46 7131029/47 7131029/48
ABEB7131028/35EA 650 DKK
ABEB7131028/35 ABEB7131028/36 ABEB7131028/37 ABEB7131028/38 ABEB7131028/39 ABEB7131028/40 ABEB7131028/41 ABEB7131028/42 ABEB7131028/43 ABEB7131028/44 ABEB7131028/45 ABEB7131028/46 ABEB7131028/47 ABEB7131028/48 ABEB7131029/35 ABEB7131029/36 ABEB7131029/37 ABEB7131029/38 ABEB7131029/39 ABEB7131029/40 ABEB7131029/41 ABEB7131029/42 ABEB7131029/43 ABEB7131029/44 ABEB7131029/45 ABEB7131029/46 ABEB7131029/47 ABEB7131029/48
ESD safety shoes, slip-on, HACCP, X-Light, 7131028/7131029
Sko og støvler
ESD safety shoe made from microfibre and a fuel resistant PU sole.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Protective steel toecap (200 J)
  • Instep area with elastic band
  • Washable at 30 °C
For use in kitchens and food industry.
Colour: White/black
Certificeringer: EN 61340 (ESD); HACCP; DGUV 112-191; EN ISO 20345:2011; S2, SRC.