VWR® Hitachi LaChromUltra II, LaChrom II, HPLC columns
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
889-0901 889-0902 889-0903 889-0904 889-0905 889-0906 889-0908 889-0909 889-0910 889-0911 889-0912
554-4078EA 4500 DKK
554-4078 554-4079 554-4080 554-4081 554-4082 554-4083 554-4085 554-4086 554-4087 554-4088 554-4089
VWR® Hitachi LaChromUltra II, LaChrom II, HPLC columns
Kromatografikolonner
Base gel: grafted material comprised of organic-inorganic silica.
Carbon content (%): approx. 20
Bonding type: polymeric
Endcapping: yes