Rods, zinc plated steel
Leverandør: R L ENTERPRISES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3109/11 3109/21 3109/31 3109/41
RLAA3109/11EA 187 DKK
RLAA3109/11 RLAA3109/21 RLAA3109/31 RLAA3109/41
Rods, zinc plated steel
Stativstænger
Zinc plated steel.
  • Domed both ends
  • Suitable for use with A shaped base (to be ordered separately)