Safety shoes, lace-up, HACCP, Anatom, 2131/2136
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2131/36 2131/37 2131/38 2131/39 2131/40 2131/41 2131/42 2131/43 2131/44 2131/45 2131/46 2131/47 2131/48 2136/36 2136/37 2136/38 2136/39 2136/40 2136/41 2136/42 2136/43 2136/44 2136/45 2136/46 2136/47 2136/48 2136/50 2136/49 2131/50 2131/49
114-7439EA 738 DKK
114-7439 114-7440 114-7441 114-7442 114-7443 114-7444 114-7445 114-7446 114-7447 114-7448 114-7449 114-7450 114-7451 114-7454 114-7455 114-7456 114-7457 114-7458 114-7459 114-7460 114-7461 114-7462 114-7463 114-7464 114-7465 114-7466 114-7468 114-7467 114-7453 114-7452
Safety shoes, lace-up, HACCP, Anatom, 2131/2136
Sko og støvler
Anti-static safety shoes made from microfibre upper and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Sanitized® treatment reduces odour and bacterial growth
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for use in kitchens.
Colour: White/black
Certificeringer: DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRC