Safety shoes, Velcro® fastening, HACCP, Anatom, 2130/2135
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2130/36 2130/37 2130/38 2130/39 2130/40 2130/41 2130/42 2130/43 2130/44 2130/45 2130/46 2130/47 2130/48 2135/36 2135/37 2135/38 2135/39 2135/40 2135/41 2135/42 2135/43 2135/44 2135/45 2135/46 2135/47 2135/48 2130/49 2130/50 2135/52 2135/51 2135/50 2135/49 2130/51 2130/52
ABEB2130/36EA 738 DKK
ABEB2130/36 ABEB2130/37 ABEB2130/38 ABEB2130/39 ABEB2130/40 ABEB2130/41 ABEB2130/42 ABEB2130/43 ABEB2130/44 ABEB2130/45 ABEB2130/46 ABEB2130/47 ABEB2130/48 ABEB2135/36 ABEB2135/37 ABEB2135/38 ABEB2135/39 ABEB2135/40 ABEB2135/41 ABEB2135/42 ABEB2135/43 ABEB2135/44 ABEB2135/45 ABEB2135/46 ABEB2135/47 ABEB2135/48 ABEB2130/49 ABEB2130/50 ABEB2135/52 ABEB2135/51 ABEB2135/50 ABEB2135/49 ABEB2130/51 ABEB2130/52
Safety shoes, Velcro® fastening, HACCP, Anatom, 2130/2135
Sko og støvler
Anti-static safety shoes made from microfibre upper and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Sanitized® treatment reduces odour and bacterial growth
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for use in kitchens.
Colour: White or black
Certificeringer: DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRC