Magnetic stirring bars sets, IKAFLON®
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4495000 4497800 4497400 4497200 4497000 4496800 4496600 4496400 4496200 4496000 4495600 4495200 4498400 4494800 4494600 4494400 4494200 4494000 4493800 4493400 4493200 4493000 4492800 4492600 4495400 4492200 4489300 4489200 4489100 4489000 4488900 4488800 4488600 4488700
IKAA4495000EA 280 DKK
IKAA4495000 IKAA4497800 IKAA4497400 IKAA4497200 IKAA4497000 IKAA4496800 IKAA4496600 IKAA4496400 IKAA4496200 IKAA4496000 IKAA4495600 IKAA4495200 IKAA4498400 IKAA4494800 IKAA4494600 IKAA4494400 IKAA4494200 IKAA4494000 IKAA4493800 IKAA4493400 IKAA4493200 IKAA4493000 IKAA4492800 IKAA4492600 IKAA4495400 IKAA4492200 IKAA4489300 IKAA4489200 IKAA4489100 IKAA4489000 IKAA4488900 IKAA4488800 IKAA4488600 IKAA4488700
Magnetic stirring bars sets, IKAFLON®
Omrørere Omrøremagneter
Glass or PTFE-coated stirring bars. Used in wide range of applications. Available in 5/set or 1/set.
  • Very good chemical resistance
  • Beaker line bars are autoclavable
  • Temperature resistant, suitable for high and low temperature applications