Water activity handheld sets, HP23-AW-A
Leverandør: ROTRONIC INSTRUMENTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HP23AWA-SET-40 HP23-AW-A
710-0995EA 32790 DKK
710-0995 620-2753
Water activity handheld sets, HP23-AW-A
Hygrometre Digitale termo-hygrometre
Water activity can be measured in production or storage rooms for the inspection of bulk materials, QC checks of cheese, meat, tobacco, building materials, animal feeds, bakery products, paper, medicines, horticulture and agriculture. The sets include advanced features for the shelf life of food, pharmaceutical and cosmetic products during storage. They document the occurrence and growth of microorganisms.
  • Battery charging function
  • Audible alarm to indicate completed measurement
  • Recalibration by the customer with the appropriate standards
  • Saves up to 20 000 data records with % RH, °C/°F, date and time
  • Quick function for fast measurement results - typically 4 to 5 minutes
  • Measurement of relative humidity, temperature and all psychrometric calculations
* Only for water activity meters i.e. HygroPalm 23-AW-A and HygroLab C1.
Bestillingsoplysninger: Please note that the probes are not included and have to be ordered separately.
Leveringsinformation: Supplied with a battery and a factory adjustment certificate.