Finnpipette™ F2 Good Laboratory Pipetting (GLP) kits
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4700870 4700880 4700885 4701070
613-2769EA 5102 DKK
613-2769 613-2770 613-2771 613-2773
Finnpipette™ F2 Good Laboratory Pipetting (GLP) kits
Pipetter Automatiske pipetter
Access everything needed for pipetting in these all-inclusive kits. The Finnpipette™ F2 GLP (Good Laboratory Pipetting) kits provide durable, high-performance Finnpipette F2 pipetters which offer maximum reliability and comfort.
  • AVG-volume gearing mechanism for accuracy and precision
  • Fully autoclavable for protection
  • Soft-touch tip ejector for light tip ejection
  • Large display for better vision ergonomics
  • Very light pipetting forces
Leveringsinformation: All kits are delivered with F-stand, Good Laboratory Pipetting guide, reagent reservoir demo pack.