Cover glasses, round, Menzel Gläser
Leverandør: Epredia
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CB00200RA120 CB00100RA120 CB00150RA120 CB00180RA120 CB00220RA120 CB00240RA120 CB00250RA133 CB00300RA140 CB00140RA120 CB00160RA120 CB00120RA120 CB00130RA020 CB00180RA220
631-1343EA 993 DKK
631-1343 631-1340 631-1341 631-1342 631-1344 631-1345 631-1346 631-1347 631-0899 630-1847 631-0713 630-1846 630-1462
Cover glasses, round, Menzel Gläser
Dækglas
These cover glasses are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles, no flaws, pre-washed
  • Made of Pure white glass of hydrolytic class 1