Ladies shorts, C.I., Design C, royal blue
Leverandør: FRISTADS KANSAS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
846530131C44 846530131C46 846530131C48 846530131C50 846530131C52 846530131C54 846530131C56 846530131C58 846530131C60 846530131C62 846530210C44 846530210C46 846530210C48 846530210C50 846530210C52 846530210C54 846530210C56 846530210C58 846530210C60 846530210C62 846530231C44 846530231C46 846530231C48 846530231C50 846530231C52 846530231C54 846530231C56 846530231C58 846530231C60 846530231C62 846530331C44 846530331C46 846530331C48 846530331C50 846530331C52 846530331C54 846530331C56 846530331C58 846530331C60 846530331C62 846530440C44 846530440C46 846530440C48 846530440C50 846530440C52 846530440C54 846530440C56 846530440C58 846530440C60 846530440C62 846530520C44 846530520C46 846530520C48 846530520C50 846530520C52 846530520C54 846530520C56 846530520C58 846530520C60 846530520C62 846530530C44 846530530C46 846530530C48 846530530C50 846530530C52 846530530C54 846530530C56 846530530C58 846530530C60 846530530C62 846530539C44 846530539C46 846530539C48 846530539C50 846530539C52 846530539C54 846530539C56 846530539C58 846530539C60 846530539C62 846530640C44 846530640C46 846530640C48 846530640C50 846530640C52 846530640C54 846530640C56 846530640C58 846530640C60 846530640C62 846530730C44 846530730C46 846530730C48 846530730C50 846530730C52 846530730C54 846530730C56 846530730C58 846530730C60 846530730C62 846530940C44 846530940C46 846530940C48 846530940C50 846530940C52 846530940C54 846530940C56 846530940C58 846530940C60 846530940C62 846530941C44 846530941C46 846530941C48 846530941C50 846530941C52 846530941C54 846530941C56 846530941C58 846530941C60 846530941C62
KWIN846530131C44EA 0 DKK
KWIN846530131C44 KWIN846530131C46 KWIN846530131C48 KWIN846530131C50 KWIN846530131C52 KWIN846530131C54 KWIN846530131C56 KWIN846530131C58 KWIN846530131C60 KWIN846530131C62 KWIN846530210C44 KWIN846530210C46 KWIN846530210C48 KWIN846530210C50 KWIN846530210C52 KWIN846530210C54 KWIN846530210C56 KWIN846530210C58 KWIN846530210C60 KWIN846530210C62 KWIN846530231C44 KWIN846530231C46 KWIN846530231C48 KWIN846530231C50 KWIN846530231C52 KWIN846530231C54 KWIN846530231C56 KWIN846530231C58 KWIN846530231C60 KWIN846530231C62 KWIN846530331C44 KWIN846530331C46 KWIN846530331C48 KWIN846530331C50 KWIN846530331C52 KWIN846530331C54 KWIN846530331C56 KWIN846530331C58 KWIN846530331C60 KWIN846530331C62 KWIN846530440C44 KWIN846530440C46 KWIN846530440C48 KWIN846530440C50 KWIN846530440C52 KWIN846530440C54 KWIN846530440C56 KWIN846530440C58 KWIN846530440C60 KWIN846530440C62 KWIN846530520C44 KWIN846530520C46 KWIN846530520C48 KWIN846530520C50 KWIN846530520C52 KWIN846530520C54 KWIN846530520C56 KWIN846530520C58 KWIN846530520C60 KWIN846530520C62 KWIN846530530C44 KWIN846530530C46 KWIN846530530C48 KWIN846530530C50 KWIN846530530C52 KWIN846530530C54 KWIN846530530C56 KWIN846530530C58 KWIN846530530C60 KWIN846530530C62 KWIN846530539C44 KWIN846530539C46 KWIN846530539C48 KWIN846530539C50 KWIN846530539C52 KWIN846530539C54 KWIN846530539C56 KWIN846530539C58 KWIN846530539C60 KWIN846530539C62 KWIN846530640C44 KWIN846530640C46 KWIN846530640C48 KWIN846530640C50 KWIN846530640C52 KWIN846530640C54 KWIN846530640C56 KWIN846530640C58 KWIN846530640C60 KWIN846530640C62 KWIN846530730C44 KWIN846530730C46 KWIN846530730C48 KWIN846530730C50 KWIN846530730C52 KWIN846530730C54 KWIN846530730C56 KWIN846530730C58 KWIN846530730C60 KWIN846530730C62 KWIN846530940C44 KWIN846530940C46 KWIN846530940C48 KWIN846530940C50 KWIN846530940C52 KWIN846530940C54 KWIN846530940C56 KWIN846530940C58 KWIN846530940C60 KWIN846530940C62 KWIN846530941C44 KWIN846530941C46 KWIN846530941C48 KWIN846530941C50 KWIN846530941C52 KWIN846530941C54 KWIN846530941C56 KWIN846530941C58 KWIN846530941C60 KWIN846530941C62
Ladies shorts, C.I., Design C, royal blue
Arbejdstøj
Short trousers made of 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring
Weight: 260 g/m²
These trousers can be customised. You can chose between 12 profile colours and 13 piping colours.