Cleanroom wipes, Anticon® White Magic™
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
495353-303 493200-529 LWWM1001
115-1849EA 9230 DKK
115-1849 CONT493200-529 CONTLWWM1001
Cleanroom wipes, Anticon® White Magic™
Papirservietter Servietter til Renrum Tør
100% interlock knit lightweight polyester wipes with sealed edges.
  • Sealed-edges for very low levels of particles and fibres
  • Patent manufacturing technology creates a wipe with excellent sorbency
  • Excellent for use in electronics applications
  • Available with Particle Attraction Technology (P.A.T.), which traps 35 times more particles
Anticon® White Magic is made from 100% continuous filament polyester with an interlock knit construction. The wipe exhibits low particle and fibre counts, and provides ultra-clean wiping for critical applications.