VWR® Q Path® Coverquick, Mounting Agents for Microscopy
Leverandør: VWR Q-Path Chemicals
Ratings:
Gennemsntilig vurdering: 5.0 (Bedømmelse)
Samlet vurdering
Gennemsntilig vurdering
Var denne bedømmelse brugbar? Ja Nej
Ja: 4 Nej: 0 
anatomie / ROUEN
Ja, jeg anbefaler
Produit conforme aux attentes du laboratoire. Très bon rapport qualité prix.
05547531.EA 392 DKK
05547531. 05547537. 05547530. 05547539. 05547528.
VWR® Q Path® Coverquick, Mounting Agents for Microscopy
Farvestoffer Mikroskopi og farvningsreagenser
A range of mounting media for manual and automatic mounting of slides. The Q Path® Coverquick range consists of four different mounting media with different viscosities to meet all microscopy needs. The number after the name indicates increasing viscosity: Q Path® Coverquick 1000 has the lower viscosity (around 260 mPaS) and Q Path® Coverquick 4000 has the higher viscosity (around 920 mPaS).
  • A complete range of mounting media for histology and cytology
  • The entire range is suitable for manual mounting
  • For automatic mounting, Q Path® Coverquick 3000 and 4000 are recommended
  • Delivered in PET amber bottle, only Q Path® Coverquick 3000 is delivered in a aluminium bottle
  • Dries quickly in 15 to 20 min