IP ActivFlo Biopsy III cassettes, loose
Leverandør: LEICA HISTOLOGY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
39LC-625-1 39LC-625-5 39LC-625-10 39LC-625-11 39LC-625-2 39LC-625-3 39LC-625-6 39LC-625-7 39LC-625-8 39LC-625-9 39LC-625-4
720-2363EA 1520 DKK
720-2363 720-2662 720-2652 720-2654 720-2656 720-2658 720-2664 720-2666 720-2668 720-2670 720-2660
IP ActivFlo Biopsy III cassettes, loose
Histologikassetter Indstøbningskassetter, Biopsiprøver
IP ActivFlo Biopsy III cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,26 mm pore size, the cassettes can secure the smallest specimens, eliminating the need for biopsy bags and sponges. The cassettes also feature sloped walls, creating a basket for small tissue management. These two piece cassettes with hinged lid have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
  • Available in loose packaging for easy printer loading
  • Choose from several colours in quantities of 500