IP ActivFlo Biopsy I cassettes, loose
Leverandør: LEICA HISTOLOGY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
39LC-605-1 39LC-605-9 39LC-605-2 39LC-605-10 39LC-605-8 39LC-605-3 39LC-605-4 39LC-605-5 39LC-605-6 39LC-605-7 39LC-605-11
720-2361EA 654 DKK
720-2361 720-2649 720-2495 720-2491 720-2647 720-2497 720-2499 720-2641 720-2643 720-2645 720-2493
IP ActivFlo Biopsy I cassettes, loose
Histologikassetter Indstøbningskassetter, Biopsiprøver
IP ActivFlo Biopsy I cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,67 mm pore size, the cassettes include an innovative V-shape pore design that pulls fluid in and forces air out. The cassettes feature sloped walls, creating a basket for small tissue management. These one piece cassettes with ‘frangible lids’ have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
  • Available in loose packaging for easy printer loading
  • Choose from several colours in quantities of 500