Capillary columns, Rtx®-Wax
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41601 41602 41603 41604 12408 12409 12420 12421 12422 12423 12424 12425 12426 12427 12435 12436 12437 12438 12439 12440 12441 12442 12443 12451 12452 12454 12455 12457 12458
568-2461EA 0 DKK
568-2461 568-2462 568-2463 568-2464 568-2217 568-2218 568-2219 568-2220 568-2221 568-2222 568-2223 568-2224 568-2225 568-2226 568-2227 568-2228 568-2229 568-2230 568-2231 568-2232 568-2233 568-2234 568-2235 568-2236 568-2237 568-2238 568-2239 568-2240 568-2241
Capillary columns, Rtx®-Wax
Kromatografikolonner
General purpose columns for FAMEs, solvents, BTEX aromatics, flavour compounds, alcohols.
Polar phase; Crossbond® Carbowax® polyethylene glycol
Temperature range: 20 to 250 °C
Equivalent to USP G14, G15, G16, G20, G39 phases.