2-Mercaptothiophene 97% (as monomer) contains dimer
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 97%
CAS No.: 7774-74-5
Molecular Formula: C4H4S2
Formula Weight: 116.20
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6170
Odour: Unpleasant
Boiling Point: 62-65°/12mm
Beilstein Reference: 104650
MDL No.: MFCD00051666
Formel:
C₄H₄S₂ Kogepunkt: 62…65 °C (12 mmHg) Massefylde: 1,247 Flammepunkt: 66 °C (151 °F) |
MDL Number:
MFCD00051666 CAS-nummer: 7774-74-5 EINECS: 231-881-1 UN nummer: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...