1-Naphthalenemethanol ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Naphthalen-1-ylmethanol

N0633-25G N0633-5G
TCIAN0633-25GEA 862 DKK
TCIAN0633-25G TCIAN0633-5G
1-Naphthalenemethanol ≥95.0% (by GC)
1-Naphthalenemethanol
Formel: C₁₁H₁₀O
Kogepunkt: 301°C
MDL Number: MFCD00004044
CAS-nummer: 4780-79-4
EINECS: 225-324-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04e4c54141a15fb393a0088a227c8b68b75e3f8c&vid=04e4c54141a15fb393a0088a227c8b68b75e3f8c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR